missing translation for 'onlineSavingsMsg'
Learn More

Sodium iodide hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11387027
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium Iodide is used in the synthesis of dye-sensitized solar cells. Also used in the preparation of propargylamines as inhibitors of lysine specific. Demethylase. Used in photography and as analytical reagents. They are used in the measurement of the energy of gamma rays, by measuring the amplitude of pulses of light generated by electrons which are excited by the gamma rays and in the Finkelstein reaction via an SN2 pathway, detection of radiation, and in the conversion of an alkyl chloride into an alkyl iodide. Sodium Iodide is used in PCR and to produce chemokines, particularly CCL2, CXCL8, and CXC14 by stimulation of thyroid follicular cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003532

Specifications

Physical Form Crystalline Aggregates
Quantity 25 g
Solubility Information Soluble in water (1840mg/ml at 25°C),alcohol,glycerol,and acetone. Insoluble in ether.
Formula Weight 149.89 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic™
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Sodium iodide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'