missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, Purum, ≥32%, Honeywell Fluka™

Product Code. 15680790
Click to view available options
Quantity:
2.5 L
5 L
35 kg
4 x 2.5 L
4 x 5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum,≥32%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1824
Quantity 35 kg
Formula Weight 40
Percent Purity >32%
Grade purum
Packaging Paper / Poly Bag
Vapor Pressure 3mmHg (37°C)
Chemical Name or Material Sodium hydroxide solution, Purum, ≥32%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'