missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N, Honeywell™

Product Code. 15653060
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

≥32%, for determination of nitrogen, ≤0.0001% N

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point 120°C
CAS Min % ≥50.0000%, ≥25.0000%
CAS Max % <75.0000%, <50.0000%
Linear Formula NaOH
UN Number UN1824
Density 1.350 g/cm3
Quantity 6 x 1 L
Formula Weight 40.0
Percent Purity >32%
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, ≥32%, for determination of nitrogen, ≤0.0001% N
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'