missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, Reagent Grade, 97%, 20 to 40 Mesh, Honeywell™
Reagent Grade, beads, 20-40 mesh, 97%
42.83 € - 448.52 €
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
MDL Number | MFCD00003548 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15665140
|
Honeywell Research Chemicals
367176-500G |
500 g | Poly Bottle | ||||||
15645140
|
Honeywell Research Chemicals
367176-2.5KG |
2.5 kg | Poly Bottle |
190.19 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15635140
|
Honeywell Research Chemicals
367176-12KG |
12 kg | Drum with Inliner | ||||||
15655140
|
Honeywell Research Chemicals
367176-25KG |
25 kg | Drum with Inliner | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
Colorless or White | |
1310-73-2 | |
HNaO | |
MFCD00003548 | |
2.13g/cm3 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40g/mol | |
Reagent | |
18mmHg (20°C), 3mmHg (37°C) | |
Sodium hydroxide |
1390°C | |
318°C | |
NaOH | |
UN1823 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
97% | |
>1 (vs air) | |
Beads |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'dotInformation' : 8/PGII
missing translation for 'rtecsNumber' : 215-185-5
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.