missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, purum p.a., ≥98% (T), ACS Reagent, Honeywell™
Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 1 kg
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Linear Formula | NaOH |
UN Number | UN1823 |
Quantity | 5 kg |
Formula Weight | 40g/mol |
Percent Purity | ≥98% (T) |
Vapor Density | >1 (vs air) |
Vapor Pressure | 3 mmHg (37°C), 18 mmHg (20°C) |
Chemical Name or Material | Sodium hydroxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'