missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals
21.40 € - 520.00 €
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10197850
|
Thermo Scientific Acros
424330250 |
25 g |
21.40 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10475091
|
Thermo Scientific Acros
424335000 |
500 g |
30.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10460394
|
Thermo Scientific Acros
424330010 |
1 kg |
41.20 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10187850
|
Thermo Scientific Acros
424330025 |
2.5 kg |
88.60 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10776232
|
Thermo Scientific Acros
424330100 |
10 kg |
254.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11964131
|
Thermo Scientific Acros
424330251 |
25 kg |
520.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1310-73-2 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Specifications
White | |
1310-73-2 | |
97+% | |
NaOH | |
15, 8761 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
ACS Reagent | |
Pellet |
1390.0°C | |
318.0°C | |
HNaO | |
01,1083; 05,616; 07,336; 08,460 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
≥97% | |
1mbar at 700°C | |
Sodium hydroxide |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Emergency Overview
Causes severe burns by all exposure routes. Hygroscopic.
NFPA
Health: 3
Flammability:
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium hydroxide
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only