missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, ≥97.0% (T), Honeywell™

Product Code. 15606830
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥97.0% (T), pellets

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Linear Formula NaOH
UN Number UN1823
Quantity 1 kg
Formula Weight 40g/mol
Percent Purity ≥97.0% (T)
Packaging Poly bottle
Vapor Density >1 (vs air)
Vapor Pressure 18mmHg (20°C), 3mmHg (37°C)
Chemical Name or Material Sodium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'