missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, 10N Solution, Biotech Reagent, J.T. Baker™
Click to view available options
Quantity:
1 L
4 L
19 L
Packaging:
Narrow Mouth Polyethylene Bottle
Poly Bottle
Specifications
Specifications
Quantity | 19 L |
Formula Weight | 40 |
Grade | Biotech Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 1.32kg |
Chemical Name or Material | Sodium Hydroxide, 10N Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'