missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide, 0.2 N standard solution, Thermo Scientific Chemicals

25.43 € - 46.73 €

Specifications

Chemical Name or Material Sodium hydroxide
Color Colorless
CAS 1310-73-2, 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10554901
View Documents
Thermo Scientific Acros
349685000
500 mL Plastic bottle
25.43 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10137983
View Documents
Thermo Scientific
349680025
2.5 L Plastic bottle
46.73 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium hydroxide, 0.2 N standard solution, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.