Learn More
Sodium hydrogen selenite, 96%, Thermo Scientific Chemicals
495.00 €
Chemical Identifiers
CAS | 7782-82-3 |
---|---|
Molecular Formula | Na2O3Se |
Molecular Weight (g/mol) | 172.95 |
MDL Number | MFCD00003525 |
InChI Key | BVTBRVFYZUCAKH-UHFFFAOYSA-L |
Synonym | sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide |
PubChem CID | 23669629 |
SMILES | [Na+].[Na+].[O-][Se]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421900
|
Thermo Scientific Alfa Aesar
A12173.0B |
1000 g | N/A | ||||||
Description
Sodium hydrogen selenite is used as a culture media additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hydrogen selenite is used as a culture media additive.
Solubility
Soluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
7782-82-3 | |
172.95 | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
23669629 |
Na2O3Se | |
MFCD00003525 | |
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide | |
[Na+].[Na+].[O-][Se]([O-])=O |
Specifications
7782-82-3 | |
MFCD00003525 | |
sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide | |
BVTBRVFYZUCAKH-UHFFFAOYSA-L | |
sodium;hydrogen selenite | |
23669629 | |
98% | |
Sodium hydrogen selenite |
Na2O3Se | |
UN2630 | |
Soluble in water. | |
[Na+].[Na+].[O-][Se]([O-])=O | |
172.95 | |
150.96 | |
Hygroscopic |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H300-H331-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENITES
missing translation for 'einecsNumber' : 231-966-3
missing translation for 'rtecsNumber' : VS7500000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only