Learn More
Sodium fumarate, 98%, Thermo Scientific Chemicals
16.90 € - 248.00 €
Chemical Identifiers
CAS | 17013-01-3 |
---|---|
Molecular Formula | C4H2Na2O4 |
Molecular Weight (g/mol) | 160.04 |
MDL Number | MFCD00064567 |
InChI Key | VXXVUHAXJHEYFH-SEPHDYHBSA-N |
Synonym | disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate |
PubChem CID | 87110070 |
IUPAC Name | (E)-but-2-enedioic acid;sodium |
SMILES | C(=CC(=O)O)C(=O)O.[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10291542
|
Thermo Scientific Acros
215530050 |
5 g | Glass bottle |
16.90 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10457192
|
Thermo Scientific Acros
215531000 |
100 g | Glass bottle |
24.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772505
|
Thermo Scientific Acros
215535000 |
500 g | Glass bottle |
69.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15139233
|
Thermo Scientific Acros
215530025 |
2.5 kg | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17013-01-3 | |
160.04 | |
VXXVUHAXJHEYFH-SEPHDYHBSA-N | |
87110070 | |
C(=CC(=O)O)C(=O)O.[Na].[Na] |
C4H2Na2O4 | |
MFCD00064567 | |
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate | |
(E)-but-2-enedioic acid;sodium |
Specifications
17013-01-3 | |
Authentic | |
C4H2Na2O4 | |
MFCD00064567 | |
10,4165 | |
Solubility in water: 228g/L water. Other solubilities: insoluble in ethanol, aceton | |
C(=CC(=O)O)C(=O)O.[Na].[Na] | |
160.04 | |
160.04 | |
Crystalline Powder |
White | |
98% | |
NaO2CCH=CHCO2Na | |
02,74 | |
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate | |
VXXVUHAXJHEYFH-SEPHDYHBSA-N | |
(E)-but-2-enedioic acid;sodium | |
87110070 | |
98% | |
Sodium fumarate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 241-087-7
RUO – Research Use Only