missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecylsulfate, ≥98%, Ultrapure, Thermo Scientific Chemicals
43.66 € - 193.98 €
Chemical Identifiers
CAS | 151-21-3 |
---|---|
Molecular Formula | C12H25NaO4S |
Molecular Weight (g/mol) | 288.38 |
MDL Number | MFCD00036175 |
InChI Key | DBMJMQXJHONAFJ-UHFFFAOYSA-M |
Synonym | Sodium lauryl sulfate |
PubChem CID | 3423265 |
ChEBI | CHEBI:8984 |
IUPAC Name | sodium;dodecyl sulfate |
SMILES | [Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Description
Chemical Identifiers
151-21-3 | |
288.38 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
3423265 | |
sodium;dodecyl sulfate |
C12H25NaO4S | |
MFCD00036175 | |
Sodium lauryl sulfate | |
CHEBI:8984 | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O |
Specifications
151-21-3 | |
∼206°C | |
CH3(CH2)11OSO3Na | |
UN2926 | |
Sodium lauryl sulfate | |
[Na+].CCCCCCCCCCCCOS([O-])(=O)=O | |
288.38 | |
CHEBI:8984 | |
≥98% | |
Sodium Dodecylsulfate |
Crystalline Flakes | |
C12H25NaO4S | |
MFCD00036175 | |
3599286 | |
DBMJMQXJHONAFJ-UHFFFAOYSA-M | |
sodium;dodecyl sulfate | |
3423265 | |
288.38 | |
Ultra Pure |
Safety and Handling
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P501c
H302-H315-H318-H500
missing translation for 'einecsNumber' : 205-788-1
missing translation for 'rtecsNumber' : WT1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only