missing translation for 'onlineSavingsMsg'
Learn More

Sodium dodecyl sulfate, 95%, MP Biomedicals™

Product Code. 11466051
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Approximately 95% alkyl sulfate, predominantly composed of C12 and C14 homologs

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-21-3
Molecular Formula C12H25NaO4S
Molecular Weight (g/mol) 288.38
MDL Number MFCD00036175
InChI Key DBMJMQXJHONAFJ-UHFFFAOYSA-M
Synonym SDS, Lauryl sulfate sodium salt
PubChem CID 3423265
ChEBI CHEBI:8984
IUPAC Name sodium dodecyl sulfate
SMILES [Na+].CCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Color White
Physical Form Granular Powder
Density >1.1 at 20°C (water = 1)
Quantity 1 kg
Assay Percent Range 0.95
Formula Weight 288.38
Chemical Name or Material Sodium dodecyl sulfate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Dodecyl Sulfate
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H300-Fatal if swallowed.
  • H311-Toxic in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P361-Take off immediately all contaminated clothing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'