Learn More
Sodium cyanide, 95+%, ACS reagent, Thermo Scientific Chemicals
23.82 € - 145.73 €
Chemical Identifiers
CAS | 143-33-9 |
---|---|
Molecular Formula | CNNa |
Molecular Weight (g/mol) | 49.01 |
MDL Number | MFCD00003523 |
InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
Synonym | sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h |
PubChem CID | 8929 |
SMILES | [Na+].[C-]#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10341361
|
Thermo Scientific Acros
424300050 |
5 g | Glass Bottle |
23.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10608044
|
Thermo Scientific Acros
424301000 |
100 g | Plastic bottle | N/A | |||||
10006020
|
Thermo Scientific Acros
424305000 |
500 g | Plastic bottle |
145.73 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
143-33-9 | |
49.01 | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
8929 |
CNNa | |
MFCD00003523 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
[Na+].[C-]#N |
Specifications
143-33-9 | |
Crystalline Powder, Flakes, and Chunks | |
CNNa | |
MFCD00003523 | |
04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185 | |
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h | |
[Na+].[C-]#N | |
49.01 | |
49 | |
ACS Reagent |
White | |
95+% | |
NaCN | |
02, IV, 50 | |
15, 8741 | |
MNWBNISUBARLIT-UHFFFAOYSA-N | |
sodium iminomethanide | |
8929 | |
≥95.0% | |
Sodium cyanide, 95+% |
Safety and Handling
GHS H Statement
May be corrosive to metals. Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Causes damage to organs through prolonged or repeated exposure. Very toxic to aquatic life with long lasting effects. Contact with acids liberates very toxic gas.
GHS P Statement
Absorb spillage to prevent material damage. Wear protective gloves/protective clothing. Rinse mouth. IF ON SKIN: Gently wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Immediately call a POISON CENTER or doctor/physician.
Danger
missing translation for 'einecsNumber' : 205-599-4
missing translation for 'rtecsNumber' : VZ7525000
missing translation for 'tsca' : TSCA
- Sodium cyanide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Substances and mixtures corrosive to metals Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H290-May be corrosive to metals.
- EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.