missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride Solution 0.1M (0.1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical™
30.90 € - 136.00 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
IUPAC Name | sodium chloride |
SMILES | [Na+].[Cl-] |
Description
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
sodium chloride |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 | |
[Na+].[Cl-] |
Specifications
7647-14-5 | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 |
Liquid | |
MFCD00003477 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium chloride | |
5234 | |
Ampoule |