Learn More
Sodium chloride, 99.99% (metals basis), Thermo Scientific Chemicals
72.96 € - 247.23 €
Chemical Identifiers
CAS | 7647-14-5 |
---|---|
Molecular Formula | ClNa |
Molecular Weight (g/mol) | 58.44 |
MDL Number | MFCD00003477 |
InChI Key | FAPWRFPIFSIZLT-UHFFFAOYSA-M |
Synonym | sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex |
PubChem CID | 5234 |
ChEBI | CHEBI:26710 |
SMILES | [Na+].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354989
|
Thermo Scientific Alfa Aesar
087605.18 |
50 g |
72.96 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344989
|
Thermo Scientific Alfa Aesar
087605.30 |
250 g |
247.23 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium chloride is widely used as a food additive, and a preservative. It is employed in the Chloralkali process for producing chlorine and sodium hydroxide, in the Solvay process for producing sodium carbonate and calcium chloride, in the Mannheim process and in the Hargreaves process for producing sodium sulfate and hydrochloric acid. It is used in the manufacture of sodium chlorate, water softening and de-icing of roadways. Further it is used in several industries like pharmaceutical, textile, dyeing, rubber food and agriculture. It is used in the processing of steel, copper, vanadium, and other metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-14-5 | |
58.44 | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
5234 | |
[Na+].[Cl-] |
ClNa | |
MFCD00003477 | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
CHEBI:26710 |
Specifications
1413°C | |
7647-14-5 | |
Crystals | |
ClNa | |
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex | |
FAPWRFPIFSIZLT-UHFFFAOYSA-M | |
sodium;chloride | |
5234 | |
58.44 | |
1.5442 | |
2.165 g/cm3 at 20°C |
801°C | |
White | |
99.99% (metals basis) | |
MFCD00003477 | |
Freely soluble in water. Slightly soluble in alcohol and ammonia. | |
[Na+].[Cl-] | |
58.44 | |
CHEBI:26710 | |
Odorless | |
Hygroscopic | |
Sodium chloride |
Safety and Handling
H303: May be harmful if swallowed.
missing translation for 'einecsNumber' : 231-598-3
missing translation for 'rtecsNumber' : VZ4725000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.