missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.)
55.98 € - 1332.45 €
Chemical Identifiers
CAS | 6132-02-1 |
---|---|
Molecular Formula | CH20Na2O13 |
Molecular Weight (g/mol) | 286.14 |
MDL Number | MFCD00149178 |
InChI Key | XYQRXRFVKUPBQN-UHFFFAOYSA-L |
Synonym | sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t |
PubChem CID | 151402 |
IUPAC Name | disodium;carbonate;decahydrate |
SMILES | C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15671290
|
Honeywell-Fluka
13414-1KG |
1 kg |
55.98 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681290
|
Honeywell-Fluka
13414-2.5KG |
2.5 kg |
89.41 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15601300
|
Honeywell-Fluka
13414-6X1KG |
6 x 1 kg |
292.09 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691290
|
Honeywell-Fluka
13414-50KG |
50 kg |
1332.45 €
50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
6132-02-1 | |
286.14 | |
XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
151402 | |
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
CH20Na2O13 | |
MFCD00149178 | |
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
disodium;carbonate;decahydrate |
Specifications
6132-02-1 | |
Na2CO3 · 10H2O | |
NONH for all modes of transport | |
XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
disodium;carbonate;decahydrate | |
151402 | |
99.5 to 100.5% (ex dried subst.) | |
Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
CH20Na2O13 | |
MFCD00149178 | |
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] | |
286.14 | |
286.14g/mol | |
puriss. |
Safety and Handling
P305 + P351 + P338
H319
missing translation for 'einecsNumber' : 207-838-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium carbonate decahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium carbonate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title