missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate Decahydrate, Extra Pure, SLR, Fisher Chemical™
49.80 € - 560.00 €
Chemical Identifiers
CAS | 6132-02-1 |
---|---|
Molecular Formula | CH20Na2O13 |
Molecular Weight (g/mol) | 286.14 |
MDL Number | 149178 |
InChI Key | XYQRXRFVKUPBQN-UHFFFAOYSA-L |
Synonym | sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t |
PubChem CID | 151402 |
IUPAC Name | disodium;carbonate;decahydrate |
SMILES | C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10785531
|
Fisher Chemical
S/2720/60 |
1 kg | Plastic powder jar | N/A | |||||
10264490
|
Fisher Chemical
S/2720/62 |
2.5 kg | Plastic powder jar | N/A | |||||
10090640
|
Fisher Chemical
S/2720/65 |
5 kg | Plastic keg |
560.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
6132-02-1 | |
286.14 | |
XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
151402 | |
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
CH20Na2O13 | |
149178 | |
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
disodium;carbonate;decahydrate |
Specifications
34°C | |
White | |
Solid | |
149178 | |
XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
disodium;carbonate;decahydrate | |
151402 |
6132-02-1 | |
11 to 12 | |
CH20Na2O13 | |
sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] | |
286.14 | |
286.01g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Sodium carbonate decahydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Sodium carbonate decahydrate, Sodium carbonate