Learn More
Sodium butyrate, 98+%, Thermo Scientific Chemicals
28.84 € - 123.62 €
Chemical Identifiers
CAS | 156-54-7 |
---|---|
Molecular Formula | C4H7NaO2 |
Molecular Weight (g/mol) | 110.09 |
MDL Number | MFCD00002816 |
InChI Key | MFBOGIVSZKQAPD-UHFFFAOYSA-M |
Synonym | sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 |
PubChem CID | 5222465 |
ChEBI | CHEBI:64103 |
SMILES | [Na+].CCCC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11423980
|
Thermo Scientific Alfa Aesar
A11079.06 |
5 g |
28.84 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438943
|
Thermo Scientific Alfa Aesar
A11079.22 |
100 g |
44.02 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448943
|
Thermo Scientific Alfa Aesar
A11079.36 |
500 g |
123.62 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Decreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDecreases calcium release from intracellular stores. Inhibits histone deacetylase.Sodium butyrate is used in the treatment of human spinal muscular atrophy patients. Further, it is used in the determination of the role of histone acetylation in chromatin structure and function.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
156-54-7 | |
110.09 | |
MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
5222465 | |
[Na+].CCCC([O-])=O |
C4H7NaO2 | |
MFCD00002816 | |
sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
CHEBI:64103 |
Specifications
156-54-7 | |
C4H7NaO2 | |
MFCD00002816 | |
Hygroscopic | |
Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in ether. | |
[Na+].CCCC([O-])=O | |
110.09 | |
CHEBI:64103 | |
≥98% |
>300°C | |
CH3(CH2)2CO2Na | |
3629439 | |
sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
sodium butanoate | |
5222465 | |
110.09 | |
Sodium butyrate |
Safety and Handling
GHS H Statement
H315-H319-H335;
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 205-857-6
missing translation for 'rtecsNumber' : ET6400000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium butyrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Butanoic acid, sodium salt
RUO – Research Use Only