missing translation for 'onlineSavingsMsg'
Learn More

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals

29.04 € - 218.09 €

Specifications

Boiling Point 130.0°C to 135.0°C
Chemical Name or Material Sodium borohydride
Name Note 12% solution in 40% aq. sodium hydroxide solution
CAS 16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Color Colorless
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10310424
View Documents
Thermo Scientific Acros
389931000
100 mL Plastic bottle
29.04 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10248893
View Documents
Thermo Scientific Acros
389932500
250 mL Plastic bottle
50.55 €
250mL
Please to purchase this item. Need a web account? Register with us today!
10413205
View Documents
Thermo Scientific
389930010
1 L Plastic bottle
112.56 €
1 L
Please to purchase this item. Need a web account? Register with us today!
11360836
View Documents
Thermo Scientific Acros
389930025
2.5 L Plastic bottle
218.09 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

130.0°C to 135.0°C
12% solution in 40% aq. sodium hydroxide solution
Colorless
BH4Na
MFCD00003518
Sodium tetrahydroborate, SBH
[BH4-].[Na+]
37.83
11 to 13%
1.4
Sodium borohydride
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Liquid
NaBH4
15, 8727
YOQDYZUWIQVZSF-UHFFFAOYSA-N
sodium boranuide
37.82
Plastic bottle
1.4000g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 241-004-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.