missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borohydride, ≥99%, Honeywell Fluka™

Product Code. 15616760
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

TraceSELECT™, for the determination of hydride formers by AAS,≥99%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16940-66-2
Molecular Formula BH4Na
Molecular Weight (g/mol) 37.83
MDL Number MFCD00003518
InChI Key YOQDYZUWIQVZSF-UHFFFAOYSA-N
IUPAC Name sodium boranuide
SMILES [BH4-].[Na+]

Specifications

Boiling Point >400°C
Melting Point >360°C
Color White
Physical Form Prill or Powder
Quantity 100 g
Linear Formula NaBH4
UN Number UN1426
Formula Weight 37.83g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Glass bottle
Vapor Pressure >0.000054 Pa
Density 1.074g/cm3
Chemical Name or Material Sodium borohydride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium borohydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium tetrahydroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'