Learn More
Sodium borohydride, 98%, granules, ca 3 mm, Thermo Scientific Chemicals
71.56 € - 231.15 €
Chemical Identifiers
CAS | 16940-66-2 |
---|---|
Molecular Formula | BH4Na |
Molecular Weight (g/mol) | 37.83 |
MDL Number | MFCD00003518 |
InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
Synonym | Sodium tetrahydroborate, SBH |
IUPAC Name | sodium boranuide |
SMILES | [BH4-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10617591
|
Thermo Scientific Acros
419471000 |
100 g | Glass bottle |
71.56 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422895
|
Thermo Scientific Acros
419475000 |
500 g | Plastic Bottle |
231.15 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
16940-66-2 | |
37.83 | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide |
BH4Na | |
MFCD00003518 | |
Sodium tetrahydroborate, SBH | |
[BH4-].[Na+] |
Specifications
360.0°C | |
White | |
97.5% min. (Iodometry) | |
NaBH4 | |
1426 | |
15, 8727 | |
Solubility in water: reacts. Other solubilities: soluble in ammonia - reacts with acids, soluble in amines, pyridine and dimethylether of, diethylene glycol, insol.in other ethers, hydrocarb., alkyl chlorides, reacts with oxidizing agents | |
[BH4-].[Na+] | |
37.83 | |
98% |
16940-66-2 | |
Granules | |
BH4Na | |
MFCD00003518 | |
01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189 | |
Sodium tetrahydroborate, SBH | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.83 | |
Sodium borohydride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
May damage fertility.
May damage the unborn child.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts viol
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
IF INHALED: Remove to fresh air and keep at rest in a position comf
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 241-004-4
- Sodium borohydride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 C
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H360FD-May damage fertility. May damage the unborn child.
- H301-Toxic if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- EUH014-Reacts violently with water.
Precautionary Statement
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.