missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bicarbonate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Product Code. 15653420
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur., powder,≥99.7%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Quantity 6 x 1 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Solubility Information Soluble in water, 50g/L
Formula Weight 84.01
Percent Purity ≥99.7%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Bicarbonate, Reag. Ph. Eur.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'