missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
96.45 € - 173.00 €
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium;hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15568134
|
Avantor J.T.Baker
3506-01 |
500 g | Poly Bottle | N/A | |||||
15578134
|
Avantor J.T.Baker
3506-05 |
2.5 kg | Poly Bottle |
173.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
sodium;hydrogen carbonate |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 | |
[Na+].OC([O-])=O |
Specifications
144-55-8 | |
CHNaO3 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
BAKER ANALYZED™ ACS Reagent | |
Sodium Bicarbonate |
Powder | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01 | |
1L = 2.20kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title