missing translation for 'onlineSavingsMsg'
Learn More

Sodium bicarbonate, Honeywell Fluka™

Product Code. 15668100
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.7%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
SMILES [Na+].OC([O-])=O

Specifications

Quantity 50 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
IUPAC Name sodium;hydrogen carbonate
Formula Weight 84.01g/mol
Percent Purity 99.7 to 100.3%
Chemical Name or Material Sodium bicarbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'