missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate, Trihydrate, Crystal, U.S.P. - F.C.C., J.T. Baker™
92.95 € - 362.00 €
Chemical Identifiers
CAS | 6131-90-4 |
---|---|
Molecular Formula | C2H9NaO5 |
Molecular Weight (g/mol) | 136.079 |
InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
PubChem CID | 23665404 |
ChEBI | CHEBI:32138 |
IUPAC Name | sodium;acetate;trihydrate |
SMILES | CC(=O)[O-].O.O.O.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15598124
|
Avantor J.T.Baker
3462-01 |
500 g | Poly Bottle |
92.95 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15508134
|
Avantor J.T.Baker
3462-05 |
2.5 kg | Poly Bottle |
362.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Chemical Identifiers
6131-90-4 | |
136.079 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
C2H9NaO5 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
Specifications
6131-90-4 | |
C2H9NaO5 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08 | |
1L = 1.45kg |
Crystals | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
FCC/USP | |
Sodium Acetate, Trihydrate |