missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate (Colorless-to-white Crystals), Fisher BioReagents™
22.73 € - 34.10 €
Chemical Identifiers
CAS | 6131-90-4 |
---|---|
Molecular Formula | C2H9NaO5 |
Molecular Weight (g/mol) | 136.079 |
InChI Key | AYRVGWHSXIMRAB-UHFFFAOYSA-M |
Synonym | sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn |
PubChem CID | 23665404 |
ChEBI | CHEBI:32138 |
IUPAC Name | sodium;acetate;trihydrate |
SMILES | CC(=O)[O-].O.O.O.[Na+] |
Description
Sodium Acetate Trihydrate (Colorless-to-white Crystals)
The trihydrate form of Sodium Acetate can be used for many of the same applications as the anhydrous form.Chemical Identifiers
6131-90-4 | |
136.079 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
C2H9NaO5 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
Specifications
58°C | |
White | |
Solid | |
CH3COONa·3H2O | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08 | |
1.45g/cm³ |
6131-90-4 | |
8 to 9.5 | |
C2H9NaO5 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
99.0 to 101.0% | |
Sodium Acetate Trihydrate |
Safety and Handling
Recommended Storage : RT