missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
12 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 2.5 kg |
Linear Formula | CH3COONa · 3H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3732037 |
Formula Weight | 136.08g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Ignition Point | 1112°F |
Chemical Name or Material | Sodium Acetate Trihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction