missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate, anhydrous, ACS, 99.0% min, Thermo Scientific Chemicals

38.19 € - 327.63 €

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11347267
View Documents
Thermo Scientific Alfa Aesar
011554.30
250 g
38.19 €
250g
Please to purchase this item. Need a web account? Register with us today!
11337267
View Documents
Thermo Scientific Alfa Aesar
011554.A1
1 kg
90.45 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11357267
View Documents
Thermo Scientific Alfa Aesar
011554.A7
5 kg
327.63 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium acetate is usually used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography and it is also a plating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
sodium acetate
C2H3NaO2
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
CHEBI:32954
[Na+].CC([O-])=O

Specifications

324°C
White
99.0% min.
MFCD00012459
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
517045
82.03
Nearly odorless
White crystalline powder
Sodium acetate, anhydrous, For ACS analysis
127-09-3
Crystalline
C2H3NaO2
3595639
Soluble in water,ethanol,methanol and hydrazine.
[Na+].CC([O-])=O
82.03
CHEBI:32954
ACS
Hygroscopic
1.528 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

H500

missing translation for 'einecsNumber' : 204-823-8

missing translation for 'rtecsNumber' : AJ4300010

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.