Learn More
Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals
42.81 € - 510.54 €
Chemical Identifiers
CAS | 127-09-3 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
IUPAC Name | sodium acetate |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11311618
|
Thermo Scientific Alfa Aesar
011005.06 |
5 g |
42.81 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391608
|
Thermo Scientific Alfa Aesar
011005.14 |
25 g |
152.76 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301618
|
Thermo Scientific Alfa Aesar
011005.22 |
100 g |
510.54 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
sodium acetate |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 | |
[Na+].CC([O-])=O |
Specifications
324°C | |
Black | |
99.997% (metals basis) | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate | |
517045 | |
82.03 | |
Hygroscopic | |
1.528 |
127-09-3 | |
Powder | |
C2H3NaO2 | |
3595639 | |
Very soluble in water. Soluble in alcohol. | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
Nearly odorless | |
White crystalline powder | |
Sodium acetate, anhydrous |
Safety and Handling
H500
missing translation for 'einecsNumber' : 204-823-8
missing translation for 'rtecsNumber' : AJ4300010
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.