Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
Brand:  Fisherbrand™ 90101019
 View more versions of this product
Product Code. 15872265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Withstands moderate warming
  • Light weight
  • Complies with ISO 4142
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 4142
Soda Lime Glass
Yes
150 mm
100 Pack
16 mm