Learn More
SOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10269-1-AP-150UL
 View more versions of this product
Product Code. 16886543
  • 410.00 € / 150µL
Explore more special offers
Description
Description
SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occurring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
Specifications
Specifications
SOD1
Polyclonal
Unconjugated
SOD1
ALS, ALS1, homodimer, IPOA, SOD, SOD1, Superoxide dismutase [Cu Zn]
Rabbit
Antigen Affinity Chromatography
RUO
20655, 24786, 6647
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P00441, P07632, P08228
SOD1
SOD1 Fusion Protein Ag0335
150 μL
Primary
Human, Mouse, Rat