missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16807-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B′) have been found for this gene.Specifications
SNRPB | |
Polyclonal | |
Unconjugated | |
SNRPB | |
COD, Sm B/B, Sm protein B/B, SmB/B, SmB/SmB′, snRNP B, SNRPB, SNRPB1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6628 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14678 | |
SNRPB | |
SNRPB Fusion Protein Ag10298 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction