Learn More
SNRPB Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16807-1-AP-150UL
 View more versions of this product
Product Code. 16806554
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The protein encoded by this gene is one of several nuclear proteins that are found in common among U1, U2, U4/U6, and U5 small ribonucleoprotein particles (snRNPs). These snRNPs are involved in pre-mRNA splicing, and the encoded protein may also play a role in pre-mRNA splicing or snRNP structure. Autoantibodies from patients with systemic lupus erythematosus frequently recognize epitopes on the encoded protein. Two transcript variants encoding different isoforms (B and B′) have been found for this gene.
Specifications
Specifications
SNRPB
Polyclonal
Unconjugated
SNRPB
COD, Sm B/B, Sm protein B/B, SmB/B, SmB/SmB′, snRNP B, SNRPB, SNRPB1
Rabbit
Antigen Affinity Chromatography
RUO
6628
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14678
SNRPB
SNRPB Fusion Protein Ag10298
150 μL