Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 10 cSt (40°C), Paragon Scientific™
Certified in strict accordance with ASTM D2162 at 40 °C and 100 °C under our ISO 17025 and ISO 17034 accreditation and manufactured specifically for the users of ASTM D7279, Standard Test Method for Kinematic Viscosity of Transparent and Opaque Liquids.
Brand:  Paragon Scientific HVS03
Product Code. 16060170
  • 43.05 € / 60mL
Explore more special offers
Description
Description
  • Fully traceable to international standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Certified in strict accordance with ASTM D2162
  • 60mL lower volume cost effective solution
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Small Sample Viscosity Standard ASTM
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : white mineral oil, distillates
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Nominal values quoted. Certified Values will vary from batch to batch.