missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Small Hub Removable Needles

Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.

Brand:  Hamilton™ 7804-03

 View more versions of this product

Product Code. 15927335

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Autoclavable
  • Standard needle length is 2"/51mm
  • Can be used with (RN)-style syringes that are 100μL and smaller
Specifications

Specifications

Autoclavable
26 G
0.46 mm
2, 3, or 4
Stainless Steel
0.26 mm
Removable Custom Needle
Small Removable Syringes (100μL and smaller)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers