Learn More
SLITRK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13835-1-AP-20UL
 View more versions of this product
Product Code. 16892224
  • 145.00 € / 20µL
Explore more special offers
Description
Description
SLIT and NTRK-like family 4 (Slitrk4) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk4 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.
Specifications
Specifications
SLITRK4
Polyclonal
Unconjugated
SLITRK4
SLIT and NTRK like protein 4, SLITRK4
Rabbit
Antigen Affinity Chromatography
RUO
139065, 245446, 302473
-20°C
Liquid
Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q810B8, Q8IW52
SLITRK4
SLITRK4 Fusion Protein Ag4781
20 μL
Primary
Mouse, Rat, Human