Learn More
SLC25A20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19363-1-AP-20UL
 View more versions of this product
Product Code. 16858964
  • 145.00 € / 20µL
Explore more special offers
Description
Description
SLC25A20 is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space.It mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants.This gene product is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space. This protein mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants.
Specifications
Specifications
SLC25A20
Polyclonal
Unconjugated
SLC25A20
CAC, CACT, SLC25A20
Rabbit
Antigen Affinity Chromatography
RUO
117035, 57279, 788
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43772, P97521, Q9Z2Z6
Slc25a20
SLC25A20 Fusion Protein Ag6896
20 μL
Primary
Rat, Human, Mouse