Learn More
SIRT2 Mouse anti-Human, CoraLite 488, Clone: 1D8G10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66410-100UL
Product Code. 16810796
  • 450.00 € / 100µL
Explore more special offers
Description
Description
The SIRT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SIRT2. This antibody recognizes human, mouse, rat, pig antigen. The SIRT2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
SIRT2
Monoclonal
CoraLite 488
100 μL
22933
IgG1
Immunofluorescence
1D8G10
Mouse
Primary
Human, Mouse, Rat, Pig