Learn More
Simport™ Scientific T100 BIOTUBE™ Tubes
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
Specifications
Packaging Strips of 8 tubes, 600/Cs
View More Specs
Description
Description
Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies.
  • Polypropylene construction
  • Gross volume: 1.2mL, with cap: 1.1mL
  • Caps are made of polyethylene
  • Used for preocedures requiring low surface tension such as protein, nucleic acid work
  • Designed to avoid potentially harmful lubricants while minimizing liquid retention
Applications:
  • Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
Specifications
Specifications
Strips of 8 tubes, 600/Cs