Learn More
Simport™ Scientific Urine Collection URISAFE™ Containers
User-friendly to both patients and laboratory personnel
Description
Description
User-friendly to both patients and laboratory personnel
  • Innovative cap features a large opening for easy collection
  • A leakproof-cap liner ensures spill-free sample transport
  • Vertical and horizontal graduations allow for accurate and easy reading, eliminating transfer to graduated cylinder for volume measurement
  • Containers can be subjected to freezing, thawing and EtO gas sterilization without causing changes in materials or physical appearance
  • Resistant to hydrochloric acid
  • Convenient refrigerator storage with less wasted space
  • Containers are available in 3L and 4L tilt size
  • The 4L bottle allows a clean and easy transfer to tubes by simply tilting the container forward without lifting