missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Simport™ Scientific Tamper Evident Self-Standing Transport Tubes
For all applications requiring tamper evident seal for utmost security and sample integrity
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Sterility | Non-sterile |
For Use With (Application) | Storage and transportation of biological materials |
Description
- For secure short and long term storage
- Provide strength and clarity
- Non-toxic, non sterile polypropylene made etched or non graduated tubes
- Exhibit unique design features
- With external threads for providing smooth and uniform inner surface
- Closure offers unique frangible band design
- Combination of tamper evident and resistant closure system
- For clinical trials and expensive reagents in diagnostic kits
- Caps sold separately
CAPS SOLD SEPARATELY
These unique transport tubes incorporate a seal that needs to be broken in order to open it, which leaves visual indication that the vial has been opened.
Specifications
Autoclavable | |
Polypropylene | |
Non-sterile | |
Storage and transportation of biological materials |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title