missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) oxide, powder, 1.0 micron, 99.9%, Thermo Scientific Chemicals

Product Code. 11393876
Click to view available options
Quantity:
25 g
100 g

Silicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon(IV) oxide is used in a variety of applications such as manufacturing of glass, ceramics and abrasives, and water filtration. It is a common additive in dentifrice oral care applications and the production of foods, where it is used primarily as an anti-caking and free-flow agent in powdered foods, or to absorb water in hygroscopic applications.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with hydrogen fluoride.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1710°C
pH 6
Physical Form Powder
Quantity 100 g
Merck Index 14,8493
Solubility Information Insoluble in water.
IUPAC Name silanedione
Formula Weight 60.09
Percent Purity 99.9%
Odor Odorless
Density 2
Chemical Name or Material Silicon(IV) oxide, 1.0 micron
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'