missing translation for 'onlineSavingsMsg'
Learn More

Silicon(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11364969
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Light, Air and Moisture Sensitive

Silicon(IV) iodide is used as a precursor to prepare silicon amides. It finds application in the manufacture and etching of silicon in microelectronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-84-4
Molecular Formula I4Si
Molecular Weight (g/mol) 535.70
MDL Number MFCD00054041
InChI Key CFTHARXEQHJSEH-UHFFFAOYSA-N
Synonym silicon tetraiodide, silane, tetraiodo, silicon iv iodide, periodosilane, tetraiodosilicon, acmc-1bu37, silicon tetraiodide trace metals basis, silicon iv iodide trace metals basis 1g
PubChem CID 83498
IUPAC Name tetraiodosilane
SMILES I[Si](I)(I)I

Specifications

Color White
Physical Form Crystals
Quantity 1 g
Assay Percent Range 99% (metals basis)
Formula Weight 535.7
Chemical Name or Material Silicon(IV) iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(IV) iodide, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.