Learn More
Silicon(IV) bromide, Thermo Scientific Chemicals
121.00 € - 452.00 €
Chemical Identifiers
CAS | 7789-66-4 |
---|---|
Molecular Formula | Br4Si |
Molecular Weight (g/mol) | 347.701 |
MDL Number | MFCD00049530 |
InChI Key | AIFMYMZGQVTROK-UHFFFAOYSA-N |
Synonym | silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj |
PubChem CID | 82247 |
IUPAC Name | tetrabromosilane |
SMILES | [Si](Br)(Br)(Br)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11381559
|
Thermo Scientific Alfa Aesar
014029.09 |
10 g |
121.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391559
|
Thermo Scientific Alfa Aesar
014029.18 |
50 g |
452.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silicon(IV) bromide is used to grow thin films of silicon by the chemical vapor deposition method. It is also used for ceramics, sealants and the production of many cutting tools.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-66-4 | |
347.701 | |
AIFMYMZGQVTROK-UHFFFAOYSA-N | |
82247 | |
[Si](Br)(Br)(Br)Br |
Br4Si | |
MFCD00049530 | |
silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj | |
tetrabromosilane |
Specifications
154°C | |
7789-66-4 | |
Liquid | |
MFCD00049530 | |
14,8499 | |
AIFMYMZGQVTROK-UHFFFAOYSA-N | |
tetrabromosilane | |
82247 | |
Stench | |
2.800 g/mL |
5°C | |
Colorless to Yellow | |
Br4Si | |
UN3264 | |
silicon iv bromide, silicon tetrabromide, silane, tetrabromo, sibr4, unii-8gac9ywl42, 8gac9ywl42, silicon bromide, acmc-20ajqj | |
[Si](Br)(Br)(Br)Br | |
347.701 | |
347.72 | |
Moisture sensitive | |
Silicon(IV) bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 232-182-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only