missing translation for 'onlineSavingsMsg'
Learn More

Silicon(II) oxide, 99.99% (metals basis), Thermo Scientific Chemicals

164.82 € - 480.39 €

Chemical Identifiers

CAS 10097-28-6
Molecular Formula OSi
Molecular Weight (g/mol) 44.084
MDL Number MFCD00151536
InChI Key LIVNPJMFVYWSIS-UHFFFAOYSA-N
Synonym silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
PubChem CID 66241
ChEBI CHEBI:30588
IUPAC Name oxoniumylidynesilanide
SMILES [O+]#[Si-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333438
SDS View Documents
Thermo Scientific Alfa Aesar
036347.14
25 g
164.82 €
25g
Please to purchase this item. Need a web account? Register with us today!
11323438
SDS View Documents
Thermo Scientific Alfa Aesar
036347.22
100 g
480.39 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A major application of silicon oxide is production of portland cement. It is used in food and pharmaceutical applications and in semiconductor industry. It is the major precursor for preparing glass and silicon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10097-28-6
44.084
LIVNPJMFVYWSIS-UHFFFAOYSA-N
66241
oxoniumylidynesilanide
OSi
MFCD00151536
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
CHEBI:30588
[O+]#[Si-]

Specifications

1880°C
10097-28-6
Lump
OSi
silicon monoxide, silylene, oxo, silicon ii oxide, unii-1oqn9cbg7l, 1oqn9cbg7l, silicon ii oxide, optical grade, sio, oxidosilicon, oxosilanylidene, oxosilylene
LIVNPJMFVYWSIS-UHFFFAOYSA-N
oxoniumylidynesilanide
66241
44.09
2.13 g/cm3 at 20°C
>1702°C
Black
99.99% (metals basis)
MFCD00151536
Insoluble in water.
[O+]#[Si-]
44.084
CHEBI:30588
Odorless
Silicon(II) oxide
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-232-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.