missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica Gel, Honeywell Fluka™
With moisture indicator free of heavy metals
68.83 € - 1167.13 €
Chemical Identifiers
CAS | 112926-00-8 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00011232 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15661360
|
Honeywell-Fluka
13767-500G |
500 g |
68.83 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15641360
|
Honeywell-Fluka
13767-1KG |
1 kg |
118.33 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15651360
|
Honeywell-Fluka
13767-2.5KG |
2.5 kg |
229.79 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691360
|
Honeywell-Fluka
13767-6X500G |
6 x 500 g |
350.09 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15671360
|
Honeywell-Fluka
13767-6X1KG |
6 x 1 kg |
599.37 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15681360
|
Honeywell-Fluka
13767-6X2.5KG |
6 x 2.5 kg |
1167.13 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
112926-00-8 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
CHEBI:30563 | |
O=[Si]=O |
O2Si | |
MFCD00011232 | |
24261 | |
dioxosilane |
Specifications
112926-00-8 | |
O2Si | |
NONH for all modes of transport | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 |
Silica Gel | |
MFCD00011232 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
Silica Gel |
Safety and Handling
missing translation for 'einecsNumber' : 231-545-4