missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silica, Active, EcoChrom™, MP Biomedicals™
Click to view available options
Quantity:
500 g
Specifications
Specifications
CAS | 7631-86-9 |
Quantity | 500 g |
Molecular Formula | O2Si |
MDL Number | MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733 |
Synonym | Silica, Dioxosilane |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
SMILES | O=[Si]=O |
IUPAC Name | dioxosilane |
Molecular Weight (g/mol) | 60.08 |
PubChem CID | 24261 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction