missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGG_07580 (Glucose Oxidase), Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against native Glucose Oxidase (Aspergillus niger).
Brand: Abnova PAB28099.100ug
This item is not returnable.
View return policy
Specifications
MGG_07580 (Glucose Oxidase) | |
Polyclonal | |
Sheep polyclonal antibody raised against native Glucose Oxidase (Aspergillus niger). | |
Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
MG07580.4 | |
Sheep | |
100 μg | |
Primary | |
Fungi | |
Lyophilized |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:5500 - 1:25000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
MGG_07580 | |
MGG_07580 | |
Native purified Aspergillus nige Glucose Oxidase. | |
RUO | |
2683500 | |
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
For Research Use Only