missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Slide Holder, 10 Slide Capacity
Store 10 slides vertically with the Epredia™ Shandon™ Slide Holder.
Brand: Epredia™ 147
This item is not returnable.
View return policy
Description
- Features individually stamped slots
- Holds 76 × 26mm
Specifications
Slide Holder | |
10 | |
26 mm |
Slides | |
76 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction